Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromoindazole-1-carboxylic acid tert-butyl ester 97%

Supplier: AMBEED, INC
Warning

A504566-10G A504566-1G A504566-250MG A504566-5G
77034-520EA 249 USD InStock
77034-520 77034-522 77034-524 77034-526
6-Bromoindazole-1-carboxylic acid tert-butyl ester 97%
6-Bromoindazole-1-carboxylic acid tert-butyl ester
Precaución: for Research Use Only


Formula: C₁₂H₁₃BrN₂O₂
MDL Number: MFCD06739133
CAS Number: 877264-77-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty