Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloro(1,3-dimesityl-1H-imidazol-2(3H)-ylidene)aurate(I) 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A504795-100MG A504795-250MG A504795-50MG A504795-1G
76991-282EA 53 USD InStock
76991-282 76991-284 76991-286 77684-324
Chloro(1,3-dimesityl-1H-imidazol-2(3H)-ylidene)aurate(I) 98%, Ambeed.Inc
Chloro(1,3-dimesityl-1H-imidazol-2(3H)-ylidene)aurate(I)
Precaución: For research use only.


Formula: C₂₁H₂₄AuClN₂
MDL Number: MFCD11656059
CAS Number: 852445-81-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty