Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Hydroxy-5-methylphenyl)boronic acid 98%

Supplier: AMBEED, INC
Warning

A506263-100MG A506263-1G A506263-250MG
77017-250EA 15 USD InStock
77017-250 77017-252 77017-254
(3-Hydroxy-5-methylphenyl)boronic acid 98%
(3-Hydroxy-5-methylphenyl)boronic acid
Precaución: for Research Use Only


Formula: C₇H₉BO₃
MW: 151.95 g/mol
MDL Number: MFCD16618932
CAS Number: 1256345-79-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty