Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[(6-Chloropyridin-3-yl)carbonyl]morpholine 98%

Supplier: AMBEED, INC
Warning

A507075-100MG A507075-10G A507075-250MG A507075-5G
77034-540EA 7 USD InStock
77034-540 77034-542 77034-544 77034-546
4-[(6-Chloropyridin-3-yl)carbonyl]morpholine 98%
4-[(6-Chloropyridin-3-yl)carbonyl]morpholine
Precaución: for Research Use Only


Formula: C₁₀H₁₁ClN₂O₂
MW: 226.66 g/mol
MDL Number: MFCD03750290
CAS Number: 64614-49-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty