Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4',4''-dimethyltriphenylamine 97%

Supplier: AMBEED, INC
Warning

A509100-1G A509100-5G
76991-338EA 21 USD InStock
76991-338 76991-340
3-Bromo-4',4''-dimethyltriphenylamine 97%
3-Bromo-4',4''-dimethyltriphenylamine
Precaución: for Research Use Only


Formula: C₂₀H₁₈BrN
MW: 352.28 g/mol
MDL Number: MFCD15144691
CAS Number: 845526-91-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty