Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-2-oxo-1,2-dihydroquinoline-3-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A510076-100MG A510076-1G A510076-250MG
77017-304EA 95 USD InStock
77017-304 77017-306 77017-308
1-Methyl-2-oxo-1,2-dihydroquinoline-3-carboxylic acid 95%
1-Methyl-2-oxo-1,2-dihydroquinoline-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₁H₉NO₃
MDL Number: MFCD08754095
CAS Number: 67984-94-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty