Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-4,5,6,7-Tetrahydro-1H-indazol-5-amine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A511997-100MG A511997-1G A511997-250MG
77017-322EA 90 USD InStock
77017-322 77017-324 77017-326
(R)-4,5,6,7-Tetrahydro-1H-indazol-5-amine hydrochloride 97%
(R)-4,5,6,7-Tetrahydro-1H-indazol-5-amine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₂ClN₃
MDL Number: MFCD18909649
CAS Number: 1291790-29-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty