Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-N,N-dimethylpyridin-2-amine 98%

Supplier: AMBEED, INC
Warning

A512574-10G A512574-1G A512574-250MG A512574-5G
77034-614EA 305 USD InStock
77034-614 77034-616 77034-618 77034-620
6-Bromo-N,N-dimethylpyridin-2-amine 98%
6-Bromo-N,N-dimethylpyridin-2-amine
Precaución: for Research Use Only


Formula: C₇H₉BrN₂
MW: 201.06 g/mol
MDL Number: MFCD09878970
CAS Number: 112575-13-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty