Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-(4-(Methylthio)phenyl)ethanamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A513368-100MG A513368-1G A513368-250MG
76991-380EA 131 USD InStock
76991-380 76991-382 76991-384
(R)-1-(4-(Methylthio)phenyl)ethanamine hydrochloride 95%
(R)-1-(4-(Methylthio)phenyl)ethanamine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₄ClNS
MDL Number: MFCD16294237
CAS Number: 1391401-54-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty