Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2-bromoisobutyrate 97%

Supplier: AMBEED, INC
Warning

A514365-100G A514365-25G A514365-500G A514365-5G
77004-784EA 15 USD InStock
77004-784 77004-786 77004-788 77004-790
tert-Butyl 2-bromoisobutyrate 97%
tert-Butyl 2-bromoisobutyrate
Precaución: for Research Use Only


Formula: C₆H₁₁BrO₂
MW: 223.12 g/mol
MDL Number: MFCD00051908
CAS Number: 23877-12-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty