Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4,5,6-Tetrahydrocyclopenta[c]pyrazole 96%

Supplier: AMBEED, INC
Warning

A520095-100MG A520095-1G A520095-250MG
76991-454EA 17 USD InStock
76991-454 76991-456 76991-458
1,4,5,6-Tetrahydrocyclopenta[c]pyrazole 96%
1,4,5,6-Tetrahydrocyclopenta[c]pyrazole
Precaución: for Research Use Only


Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD05663424
CAS Number: 2214-03-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty