Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indole-4-carbohydrazide 95%

Supplier: AMBEED, INC
Warning

A522921-100MG A522921-1G A522921-250MG
76991-474EA 21 USD InStock
76991-474 76991-476 76991-478
1H-Indole-4-carbohydrazide 95%
1H-Indole-4-carbohydrazide
Precaución: for Research Use Only


Formula: C₉H₉N₃O
MW: 175.187 g/mol
MDL Number: MFCD06738885
CAS Number: 885272-22-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty