Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(tert-Butyl)-6-chloro-N-methoxy-N-methylbenzo[d]oxazole-7-sulfonamide 97%

Supplier: AMBEED, INC
Danger

A523557-1G A523557-250MG
77017-412EA 50 USD InStock
77017-412 77017-414
2-(tert-Butyl)-6-chloro-N-methoxy-N-methylbenzo[d]oxazole-7-sulfonamide 97%
2-(tert-Butyl)-6-chloro-N-methoxy-N-methylbenzo[d]oxazole-7-sulfonamide
Precaución: for Research Use Only


Formula: C₁₃H₁₇ClN₂O₄S
MW: 332.80 g/mol
MDL Number: MFCD27923096
CAS Number: 1206896-20-9
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty