Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-amino-5-bromo-1-benzofuran-2-carboxylate 98%

Supplier: AMBEED, INC
Warning

A524228-100MG A524228-1G A524228-250MG
77004-862EA 39 USD InStock
77004-862 77004-864 77004-866
Ethyl-3-amino-5-bromo-1-benzofuran-2-carboxylate 98%
Ethyl-3-amino-5-bromo-1-benzofuran-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₀BrNO₃
MDL Number: MFCD00454292
CAS Number: 330555-71-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty