Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Bromoethyl)-1H-imidazole hydrobromide 97%

Supplier: AMBEED, INC
Danger

A526573-100MG A526573-250MG
77004-894EA 128 USD InStock
77004-894 77004-896
1-(2-Bromoethyl)-1H-imidazole hydrobromide 97%
1-(2-Bromoethyl)-1H-imidazole hydrobromide
Precaución: for Research Use Only


Formula: C₅H₈Br₂N₂
MDL Number: MFCD16039341
CAS Number: 94614-83-2
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty