Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate 98%

Supplier: AMBEED, INC
Warning

A533574-100G A533574-10G A533574-1G A533574-25G A533574-5G
77017-506EA 67 USD InStock
77017-506 77017-508 77017-510 77017-512 77017-514
tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate 98%
tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₂₁NO₃
MW: 215.293 g/mol
MDL Number: MFCD04114966
CAS Number: 157634-00-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty