Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-(1-(Pyrrolidin-1-yl)ethyl)phenyl)boronic acid 95+%

Supplier: AMBEED, INC
Danger

A534083-100MG A534083-1G A534083-250MG
77017-516EA 53 USD InStock
77017-516 77017-518 77017-520
(4-(1-(Pyrrolidin-1-yl)ethyl)phenyl)boronic acid 95+%
(4-(1-(Pyrrolidin-1-yl)ethyl)phenyl)boronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₈BNO₂
MW: 219.08 g/mol
MDL Number: MFCD08059976
CAS Number: 868395-81-7
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty