Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-3-fluoro-D-phenylalanine 98%

Supplier: AMBEED, INC
Warning

A537387-10G A537387-1G A537387-25G A537387-5G
77004-990EA 51 USD InStock
77004-990 77004-992 77004-994 77004-996
Fmoc-3-fluoro-D-phenylalanine 98%
Fmoc-3-fluoro-D-phenylalanine
Precaución: for Research Use Only


Formula: C₂₄H₂₀FNO₄
MW: 405.43 g/mol
MDL Number: MFCD00672554
CAS Number: 198545-72-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty