Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,2-Trifluoroacetophenone-4-boronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A544660-1G A544660-250MG A544660-5G
76991-614EA 166 USD InStock
76991-614 76991-616 76991-618
2,2,2-Trifluoroacetophenone-4-boronic acid pinacol ester 97%
2,2,2-Trifluoroacetophenone-4-boronic acid pinacol ester


Formula: C₁₄H₁₆BF₃O₃
MDL Number: MFCD16628195
CAS Number: 1004294-77-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty