Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Carboxyethyl)-4,6-dichloro-1H-indole-2-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A545278-10MG A545278-1MG A545278-25MG A545278-5MG
77017-624EA 36 USD InStock
77017-624 77017-626 77017-628 77017-630
3-(2-Carboxyethyl)-4,6-dichloro-1H-indole-2-carboxylic acid 98%
3-(2-Carboxyethyl)-4,6-dichloro-1H-indole-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₂H₉Cl₂NO₄
MDL Number: MFCD00897722
CAS Number: 130798-51-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty