Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-[(ethylamino)methyl]phenol 97%

Supplier: AMBEED, INC
Warning

A547976-250MG A547976-5G
77017-638EA 11 USD InStock
77017-638 77017-640
4-Bromo-2-[(ethylamino)methyl]phenol 97%
4-Bromo-2-[(ethylamino)methyl]phenol
Precaución: for Research Use Only


Formula: C₉H₁₂BrNO
MW: 230.10 g/mol
MDL Number: MFCD11940289
CAS Number: 42313-78-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty