Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzyloxy)-6-fluoro-1H-indole 95%

Supplier: AMBEED, INC
Warning

A549560-100MG A549560-250MG
76991-678EA 197 USD InStock
76991-678 76991-680
4-(Benzyloxy)-6-fluoro-1H-indole 95%
4-(Benzyloxy)-6-fluoro-1H-indole
Precaución: for Research Use Only


Formula: C₁₅H₁₂FNO
MW: 241.26 g/mol
MDL Number: MFCD12405197
CAS Number: 312314-26-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty