Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Bromophenyl)-1H-pyrazole 96%

Supplier: AMBEED, INC
Danger

A552412-10G A552412-1G A552412-25G A552412-5G
77034-844EA 47 USD InStock
77034-844 77034-846 77034-848 77034-850
3-(4-Bromophenyl)-1H-pyrazole 96%
3-(4-Bromophenyl)-1H-pyrazole
Precaución: for Research Use Only


Formula: C₉H₇BrN₂
MW: 223.08 g/mol
MDL Number: MFCD01940434
CAS Number: 73387-46-9
UN: 2923
ADR: 8,6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty