Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Bromo-phenyl)-1H-pyrazole 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A553941-1G A553941-25G A553941-5G
76991-706EA 39 USD InStock
76991-706 76991-708 77684-398
1-(4-Bromo-phenyl)-1H-pyrazole 98%, Ambeed.Inc
1-(4-Bromo-phenyl)-1H-pyrazole
Precaución: For research use only.


Formula: C₉H₇BrN₂
MW: 223.08 g/mol
MDL Number: MFCD08059318
CAS Number: 13788-92-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty