Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A554214-100MG A554214-250MG
77034-868EA 16 USD InStock
77034-868 77034-870
tert-Butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate 97%
tert-Butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate
Precaución: For research use only.


Formula: C₁₁H₂₂N₂O₃
MDL Number: MFCD13152268
CAS Number: 1188265-73-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty