Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-N-(2,6-dimethylphenyl)acetamide 98%

Supplier: AMBEED, INC
Warning

A554393-100G A554393-25G A554393-500G A554393-5G
77005-104EA 53 USD InStock
77005-104 77005-106 77005-108 77005-110
2-Chloro-N-(2,6-dimethylphenyl)acetamide 98%
2-Chloro-N-(2,6-dimethylphenyl)acetamide
Precaución: for Research Use Only


Formula: C₁₀H₁₂ClNO
MDL Number: MFCD00000926
CAS Number: 1131-01-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty