Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-bromo-3-oxobutanoate 90%

Supplier: AMBEED, INC
Danger

A556966-10G A556966-1G A556966-25G A556966-5G
77005-128EA 97 USD InStock
77005-128 77005-130 77005-132 77005-134
Ethyl-4-bromo-3-oxobutanoate 90%
Ethyl-4-bromo-3-oxobutanoate
Precaución: for Research Use Only


Formula: C₆H₉BrO₃
MDL Number: MFCD07783015
CAS Number: 13176-46-0
UN: 1760
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty