Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(2,4-Dichlorophenyl)-1,3,4-oxadiazole-2-thiol 97%

Supplier: AMBEED, INC
Warning

A561629-1G A561629-250MG A561629-5G
77005-180EA 39 USD InStock
77005-180 77005-182 77005-184
5-(2,4-Dichlorophenyl)-1,3,4-oxadiazole-2-thiol 97%
5-(2,4-Dichlorophenyl)-1,3,4-oxadiazole-2-thiol
Precaución: for Research Use Only


Formula: C₈H₄Cl₂N₂OS
MDL Number: MFCD00462873
CAS Number: 23288-92-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty