Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-N4-methyl-4,5-pyrimidinediamine 98%

Supplier: AMBEED, INC
Warning

A562151-100MG A562151-1G A562151-250MG A562151-5G
77034-920EA 12 USD InStock
77034-920 77034-922 77034-924 77034-926
6-Chloro-N4-methyl-4,5-pyrimidinediamine 98%
6-Chloro-N4-methyl-4,5-pyrimidinediamine
Precaución: for Research Use Only


Formula: C₅H₇ClN₄
MDL Number: MFCD07364174
CAS Number: 52602-68-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty