Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxypropionic acid 97%

Supplier: AMBEED, INC
Warning

A562543-10G A562543-1G A562543-25G A562543-5G
77005-204EA 16 USD InStock
77005-204 77005-206 77005-208 77005-210
2-Methoxypropionic acid 97%
2-Methoxypropionic acid
Precaución: for Research Use Only


Formula: C₄H₈O₃
MDL Number: MFCD00126928
CAS Number: 4324-37-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty