Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Aminoethyl)azetidine hydrochloride, 3-BOC protected 98%

Supplier: AMBEED, INC
Warning

A566921-100MG A566921-250MG
77017-772EA 145 USD InStock
77017-772 77017-774
3-(2-Aminoethyl)azetidine hydrochloride, 3-BOC protected 98%
3-(2-Aminoethyl)azetidine hydrochloride, 3-BOC protected
Precaución: for Research Use Only


Formula: C₁₀H₂₀N₂O₂·ClH
MDL Number: MFCD09998720
CAS Number: 1170905-43-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty