Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-3-butenoate 95%

Supplier: AMBEED, INC
Danger

A567615-1G A567615-25G A567615-5G
77034-948EA 9 USD InStock
77034-948 77034-950 77034-952
tert-Butyl-3-butenoate 95%
tert-Butyl-3-butenoate
Precaución: for Research Use Only


Formula: C₈H₁₄O₂
MW: 142.20 g/mol
MDL Number: MFCD06411714
CAS Number: 14036-55-6
UN: 3272
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty