Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-5-phenylisoxazole 97%

Supplier: AMBEED, INC
Warning

A567720-10G A567720-1G A567720-250MG A567720-25G A567720-5G
77017-790EA 154 USD InStock
77017-790 77017-792 77017-794 77017-796 77017-798
3-Methyl-5-phenylisoxazole 97%
3-Methyl-5-phenylisoxazole
Precaución: for Research Use Only


Formula: C₁₀H₉NO
MDL Number: MFCD00020810
CAS Number: 1008-75-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty