Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Bromo-4'-fluoroacetanilide 98%

Supplier: AMBEED, INC
Warning

A570021-1G A570021-250MG A570021-5G
77017-802EA 28 USD InStock
77017-802 77017-804 77017-806
3'-Bromo-4'-fluoroacetanilide 98%
3'-Bromo-4'-fluoroacetanilide
Precaución: for Research Use Only


Formula: C₈H₇BrFNO
MDL Number: MFCD18071051
CAS Number: 1009-75-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty