Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[(Boc-amino)methyl]phenylboronic Acid Pinacol Ester 97%

Supplier: AMBEED, INC
Warning

A574374-100MG A574374-1G A574374-250MG
76991-836EA 51 USD InStock
76991-836 76991-838 76991-840
2-[(Boc-amino)methyl]phenylboronic Acid Pinacol Ester 97%
2-[(Boc-amino)methyl]phenylboronic Acid Pinacol Ester
Precaución: for Research Use Only


Formula: C₁₈H₂₈BNO₄
MDL Number: MFCD07371552
CAS Number: 905300-76-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty