Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-5-methylpyridine 95%

Supplier: AMBEED, INC
Warning

A579254-100MG A579254-1G A579254-250MG A579254-5G
77017-854EA 18 USD InStock
77017-854 77017-856 77017-858 77017-860
4-Bromo-2-fluoro-5-methylpyridine 95%
4-Bromo-2-fluoro-5-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD16609965
CAS Number: 1227577-02-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty