Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3-bromoindazole-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A580593-100MG A580593-250MG A580593-5G
77017-866EA 31 USD InStock
77017-866 77017-868 77017-870
tert-Butyl 3-bromoindazole-1-carboxylate 97%
tert-Butyl 3-bromoindazole-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₃BrN₂O₂
MDL Number: MFCD22380231
CAS Number: 1257296-40-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty