Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ferroceneboronic acid 95%

Supplier: AMBEED, INC
Warning

A584870-1G A584870-250MG A584870-5G
77017-910EA 35 USD InStock
77017-910 77017-912 77017-914
Ferroceneboronic acid 95%
Ferroceneboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₁BFeO₂
MDL Number: MFCD00059074
CAS Number: 12152-94-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty