Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2,3-diaminopyridine 97%

Supplier: AMBEED, INC
Warning

A585000-100MG A585000-10G A585000-1G A585000-250MG A585000-25G A585000-5G
77035-026EA 7 USD InStock
77035-026 77035-028 77035-030 77035-032 77035-034 77035-036
6-Bromo-2,3-diaminopyridine 97%
6-Bromo-2,3-diaminopyridine
Precaución: for Research Use Only


Formula: C₅H₆BrN₃
MDL Number: MFCD10697695
CAS Number: 129012-04-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty