Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-((4-methoxybenzyl)oxy)pyrimidine 95%

Supplier: AMBEED, INC
Warning

A585837-1G A585837-250MG A585837-5G
76991-954EA 101 USD InStock
76991-954 76991-956 76991-958
5-Bromo-2-((4-methoxybenzyl)oxy)pyrimidine 95%
5-Bromo-2-((4-methoxybenzyl)oxy)pyrimidine
Precaución: for Research Use Only


Formula: C₁₂H₁₁BrN₂O₂
MDL Number: MFCD12026110
CAS Number: 1159000-88-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty