Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-tert-Butoxy-4-pyridineboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A590960-1G A590960-250MG A590960-5G
76992-004EA 207 USD InStock
76992-004 76992-006 76992-008
2-tert-Butoxy-4-pyridineboronic acid pinacol ester 98%
2-tert-Butoxy-4-pyridineboronic acid pinacol ester


Formula: C₁₅H₂₄BNO₃
MDL Number: MFCD11223286
CAS Number: 1256358-89-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty