Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-Bromofuran-2-yl)(pyrrolidin-1-yl)methanone 95%

Supplier: AMBEED, INC

A601638-1G A601638-250MG A601638-25G A601638-5G
77017-982EA 35 USD InStock
77017-982 77017-984 77017-986 77017-988
(5-Bromofuran-2-yl)(pyrrolidin-1-yl)methanone 95%
(5-Bromofuran-2-yl)(pyrrolidin-1-yl)methanone
Precaución: for Research Use Only


Formula: C₉H₁₀BrNO₂
MDL Number: MFCD00460935
CAS Number: 157642-10-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty