Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GDC-0449 98%

Supplier: AMBEED, INC
Warning

A603143-1MG A603143-250MG
77035-118EA 15.92 USD InStock
77035-118 77035-120 77035-122
GDC-0449 98%
GDC-0449
Precaución: for Research Use Only


Formula: C₁₉H₁₄Cl₂N₂O₃S
MDL Number: MFCD12407408
CAS Number: 879085-55-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty