Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium (2,4-dimethylphenyl)trifluoroborate 97%

Supplier: AMBEED, INC
Warning

A604242-10G A604242-1G A604242-5G
77035-126EA 234 USD InStock
77035-126 77035-128 77035-130
Potassium (2,4-dimethylphenyl)trifluoroborate 97%
Potassium (2,4-dimethylphenyl)trifluoroborate
Precaución: for Research Use Only


Formula: C₈H₉BF₃K
MW: 212.06 g/mol
MDL Number: MFCD07368353
CAS Number: 850245-50-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty