Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-piperidine-4-carbonitrile 96%

Supplier: AMBEED, INC
Danger

A604725-1G A604725-25G
77018-004EA 40 USD InStock
77018-004 77018-006 77018-008
1-Methyl-piperidine-4-carbonitrile 96%
1-Methyl-piperidine-4-carbonitrile
Precaución: for Research Use Only


Formula: C₇H₁₂N₂
MW: 124.184 g/mol
MDL Number: MFCD06658487
CAS Number: 20691-92-3
UN: 3276
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty