Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3R,5S)-rel-3,5-Dimethylmorpholine 98%

Supplier: AMBEED, INC
Danger

A605643-1G A605643-250MG
77035-136EA 100 USD InStock
77035-136 77035-138
(3R,5S)-rel-3,5-Dimethylmorpholine 98%
(3R,5S)-rel-3,5-Dimethylmorpholine
Precaución: for Research Use Only


Formula: C₆H₁₃NO
MDL Number: MFCD12405010
CAS Number: 45597-00-0
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty