Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopropylhydrazine dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A608957-1G A608957-250MG A608957-5G
77018-034EA 39 USD InStock
77018-034 77018-036 77018-038
Cyclopropylhydrazine dihydrochloride 95%
Cyclopropylhydrazine dihydrochloride
Precaución: for Research Use Only


Formula: C₃H₁₀Cl₂N₂
MDL Number: MFCD22380553
CAS Number: 1374652-23-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty