Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodo-2-furaldehyde 98%

Supplier: AMBEED, INC
Warning

A609878-1G A609878-25G A609878-5G
76992-124EA 11 USD InStock
76992-124 76992-126 76992-128
5-Iodo-2-furaldehyde 98%
5-Iodo-2-furaldehyde
Precaución: for Research Use Only


Formula: C₅H₃IO₂
MDL Number: MFCD00159503
CAS Number: 2689-65-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty