Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fluoranthene-3-boronic acid 95%

Supplier: AMBEED, INC
Warning

A613438-250MG A613438-25G A613438-5G
76992-162EA 14 USD InStock
76992-162 76992-164 76992-166
Fluoranthene-3-boronic acid 95%
Fluoranthene-3-boronic acid
Precaución: for Research Use Only


Formula: C₁₆H₁₁BO₂
MDL Number: MFCD16660041
CAS Number: 359012-63-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty