Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylquinoline-6-carbonitrile 97%

Supplier: AMBEED, INC
Warning

A613617-100MG A613617-1G A613617-250MG
77018-058EA 28 USD InStock
77018-058 77018-060 77018-062
2-Methylquinoline-6-carbonitrile 97%
2-Methylquinoline-6-carbonitrile
Precaución: for Research Use Only


Formula: C₁₁H₈N₂
MDL Number: MFCD11520855
CAS Number: 73013-69-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty